Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(4-Bromophenyl)-5-phenylthiophene 98.0+%, TCI America™
SDP

Catalog No. B54911G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B54911G 1 g
B5491200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B54911G Supplier TCI America Supplier No. B54911G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 118621-30-0
Molecular Formula C16H11BrS
Molecular Weight (g/mol) 315.228
MDL Number MFCD21648452
InChI Key YMDCOEOZWHXGTM-UHFFFAOYSA-N
PubChem CID 10313583
IUPAC Name 2-(4-bromophenyl)-5-phenylthiophene
SMILES C1=CC=C(C=C1)C2=CC=C(S2)C3=CC=C(C=C3)Br

Specifications

Melting Point 190°C
Quantity 1 g
Formula Weight 315.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(4-Bromophenyl)-5-phenylthiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.