Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(4-Bromophenyl)benzothiazole 98.0+%, TCI America™
SDP

Catalog No. B267925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B267925G 25 g
B26795G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B267925G Supplier TCI America Supplier No. B267925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 19654-19-4
Molecular Formula C13H8BrNS
Molecular Weight (g/mol) 290.18
MDL Number MFCD00579199
InChI Key FQIRBKKYMJKENC-UHFFFAOYSA-N
Synonym 1-(2-Benzothiazolyl)-4-bromobenzene
PubChem CID 2320602
IUPAC Name 2-(4-bromophenyl)-1,3-benzothiazole
SMILES BrC1=CC=C(C=C1)C1=NC2=CC=CC=C2S1

Specifications

Melting Point 134°C
Quantity 25 g
Formula Weight 290.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(4-Bromophenyl)benzothiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.