Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(4-Chlorophenyl)ethanol 98.0+%, TCI America™
SDP

Catalog No. C223325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C223325G 25 g
C22335G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C223325G Supplier TCI America Supplier No. C223325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1875-88-3
Molecular Formula C8H9ClO
Molecular Weight (g/mol) 156.609
MDL Number MFCD00002899
InChI Key HZFRKZWBVUJYDA-UHFFFAOYSA-N
Synonym 2-4-chlorophenyl ethanol, 4-chlorophenethylalcohol, 4-chlorophenethyl alcohol, 2-4-chlorophenyl ethan-1-ol, benzeneethanol, 4-chloro, p-chlorophenethylic alcohol, 4-chlorophenethanol, 4-chlorophenyl methyl carbinol, 4-chlorobenzeneethanol, p-chlorophenethylalcohol
PubChem CID 74647
IUPAC Name 2-(4-chlorophenyl)ethanol
SMILES C1=CC(=CC=C1CCO)Cl

Specifications

Color Yellow
Quantity 25 g
Physical Form Clear Liquid at 20°C
Boiling Point 153°C
Formula Weight 156.61
Percent Purity ≥98.0% (GC)
Chemical Name or Material 2-(4-Chlorophenyl)ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.