Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Di-tert-butylphenol, 97%

Catalog No. p-3757464
Click to view available options
Quantity:
100 g
500 g
2.5 kg
25 kg
Packaging:
Glass bottle
Metal Drum

CAS: 96-76-4 | C14H22O | 206.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-76-4
MDL Number MFCD00008828
InChI Key ICKWICRCANNIBI-UHFFFAOYSA-N
Synonym 2,4-di-tert-butylphenol, 2,4-di-t-butylphenol, phenol, 2,4-bis 1,1-dimethylethyl, antioxidant no. 33, 1-hydroxy-2,4-di-tert-butylbenzene, phenol, 2,4-di-tert-butyl, 2,4-bis tert-butyl phenol, prodox 146, prodox 146a-85x, unii-fob94g6hzt
PubChem CID 7311
IUPAC Name 2,4-ditert-butylphenol
SMILES CC(C)(C)C1=CC(=C(C=C1)O)C(C)(C)C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 54.0°C to 59.0°C
Color White to Yellow
Boiling Point 264.0°C
Flash Point 115°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C14H22O
Linear Formula [(CH3)3C]2C6H3OH
Quantity 100 g
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in methanol and ether
Molecular Weight (g/mol) 206.33
Formula Weight 206.33
Percent Purity 97%
Physical Form Crystalline Solid
Chemical Name or Material 2,4-Di-tert-butylphenol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Very toxic to aquatic life with long lasting effects.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear eye protection/face protection.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 202-532-

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.