Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Diamino-6-butylamino-1,3,5-triazine 97.0+%, TCI America™
SDP

Catalog No. B072725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B072725G 25 g
B07275G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B072725G Supplier TCI America Supplier No. B072725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5606-24-6
Molecular Formula C7H14N6
Molecular Weight (g/mol) 182.23
MDL Number MFCD00059764
InChI Key CVKGSDYWCFQOKU-UHFFFAOYSA-N
Synonym 2-Butylamino-4,6-diamino-1,3,5-triazine, N2-Butylmelamine
PubChem CID 197831
IUPAC Name N2-butyl-1,3,5-triazine-2,4,6-triamine
SMILES CCCCNC1=NC(N)=NC(N)=N1

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 182.23
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Diamino-6-butylamino-1,3,5-triazine
RTECSNumber XY5852000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.