Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Diamino-6-phenyl-s-triazine ≥99%, Thermo Scientific™

Catalog No. AC406405000
Click to view available options
:
500g

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 91-76-9
CAS Min % 99.0
CAS Max % 100.0
Density 1.4000g/mL
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Molecular Formula C9H9N5
Quantity 500g
MDL Number MFCD00023187
Beilstein 26, IV, 1244
Merck Index 15, 1091
Synonym benzoguanamine, 2,4-diamino-6-phenyl-1,3,5-triazine, benzoguanimine, 1,3,5-triazine-2,4-diamine, 6-phenyl, usaf rh-5, 2,4-diamino-6-phenyl-s-triazine, 2-phenyl-4,6-diamino-s-triazine, 4,6-diamino-2-phenyl-s-triazine, benzoguanamine van, s-triazine, 2,4-diamino-6-phenyl
InChI Key GZVHEAJQGPRDLQ-UHFFFAOYSA-N
SMILES NC1=NC(=NC(N)=N1)C1=CC=CC=C1
IUPAC Name 6-phenyl-1,3,5-triazine-2,4-diamine
Molecular Weight (g/mol) 187.21
PubChem CID 7064
Formula Weight 187.2
Percent Purity ≥99.0%
Chemical Name or Material 2, 4-Diamino-6-phenyl-s-triazine, 99+%
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
Harmful to aquatic life with long lasting effects.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 202-095-6
RTECSNumber XY7000000
TSCA TSCA
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.