Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Diaminophenol dihydrochloride, 98%

Catalog No. p-3757408
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle

CAS: 137-09-7 | C6H10Cl2N2O | 197.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 137-09-7
Molecular Formula C6H10Cl2N2O
Molecular Weight (g/mol) 197.06
MDL Number MFCD00012979
InChI Key KQEIJFWAXDQUPR-UHFFFAOYSA-N
Synonym 2,4-diaminophenol dihydrochloride, amidol, dianol, acrol, 2,4-diaminophenol hcl, 2,4-diaminophenol.2hcl, phenol, 2,4-diamino-, dihydrochloride, 2,4-diaminophenol hydrochloride, diamidophenol hydrochloride, unii-t1iyy101nx
PubChem CID 8715
IUPAC Name 2,4-diaminophenol;dihydrochloride
SMILES [H+].[H+].[Cl-].[Cl-].NC1=CC=C(O)C(N)=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 222°C
Color Beige to Gray-Green
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Packaging Glass bottle
Linear Formula (H2N)2C6H3OH·2HCl
Quantity 25 g
Beilstein 13, 550
Merck Index 14, 2983
Solubility Information Solubility in water: 27.5g/100mL (15°C). Other solubilities: slightly soluble in alcohol and ether
Formula Weight 197.07
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2, 4-Diaminophenol dihydrochloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Causes serious eye damage.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 205-279-4

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.