Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dibromo-6-nitroaniline, 99%

Catalog No. aal2033306
Click to view available options
Quantity:
5 g
25 g

CAS: 827-23-6 | C6H4Br2N2O2 | 295.918 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 827-23-6
Molecular Formula C6H4Br2N2O2
Molecular Weight (g/mol) 295.918
MDL Number MFCD00012107
InChI Key OBTUHOVIVLDLLD-UHFFFAOYSA-N
Synonym benzenamine, 2,4-dibromo-6-nitro, 2,4-dibromo-6-nitrophenylamine, 2,4-dibromo-6-nitrobenzenamine, acmc-209pq0, 2-nitro-4,6-dibromoaniline, 4,6-dibromo-2-nitroaniline, 2,4-dibromo-6-nitro-aniline, 2,4-dibromo-6-nitro-phenylamine, aniline, 2,4-dibromo-6-nitro, benzenamine,2,4-dibromo-6-nitro
PubChem CID 70009
IUPAC Name 2,4-dibromo-6-nitroaniline
SMILES C1=C(C=C(C(=C1Br)N)[N+](=O)[O-])Br

Specifications

Melting Point 129°C to 131°C
Quantity 5 g
Beilstein 2805574
Formula Weight 295.92
Percent Purity 99%
Chemical Name or Material 2,4-Dibromo-6-nitroaniline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H373-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.