Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,4-Dibromobenzyl alcohol, 98%

Click to view available options
Quantity:
250mg
1g
5g
666747-06-4
C7H6Br2O
265.932
MFCD09996906
VXEVXBMLHQPKGA-UHFFFAOYSA-N
2,4-dibromophenyl methanol,2,4-dibromobenzyl alcohol,benzenemethanol, 2,4-dibromo,2,4-dibromobenzenemethanol,benzenemethanol,2,4-dibromo,pubchem9781,2,4-dibromobenzylalcohol,acmc-1bfu5,2,4-bis bromanyl phenyl methanol
17836195
(2,4-dibromophenyl)methanol
C1=CC(=C(C=C1Br)Br)CO
666747-06-4
C7H6Br2O
265.932
MFCD09996906
VXEVXBMLHQPKGA-UHFFFAOYSA-N
2,4-dibromophenyl methanol,2,4-dibromobenzyl alcohol,benzenemethanol, 2,4-dibromo,2,4-dibromobenzenemethanol,benzenemethanol,2,4-dibromo,pubchem9781,2,4-dibromobenzylalcohol,acmc-1bfu5,2,4-bis bromanyl phenyl methanol
17836195
(2,4-dibromophenyl)methanol
C1=CC(=C(C=C1Br)Br)CO
Applications
It is an halogenated compound for proteomics research. Also used as a intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Quantity 1g
Beilstein 3241294
Formula Weight 265.98
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 2,4-Dibromobenzyl alcohol

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning