Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dibromophenol, 98%

Catalog No. p-7026981
Click to view available options
Quantity:
25 g
100 g

CAS: 615-58-7 | C6H4Br2O | 251.905 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 615-58-7
Molecular Formula C6H4Br2O
Molecular Weight (g/mol) 251.905
MDL Number MFCD00002149
InChI Key FAXWFCTVSHEODL-UHFFFAOYSA-N
Synonym phenol, 2,4-dibromo, 2,4-dibromo-phenol, 2,4-dibromphenol, unii-ia75t5c9tg, 2,4-bis bromanyl phenol, ia75t5c9tg, phenol,4-dibromo, emery 9331, acmc-1bga9, 2,4-dibromophenol
PubChem CID 12005
ChEBI CHEBI:34238
IUPAC Name 2,4-dibromophenol
SMILES C1=CC(=C(C=C1Br)Br)O

Specifications

Melting Point 35°C to 38°C
Boiling Point 239°C
Flash Point >110°C (230°F)
Odor Phenol-like
Quantity 25 g
UN Number UN2811
Beilstein 1861291
Formula Weight 251.92
Percent Purity 98%
Chemical Name or Material 2,4-Dibromophenol
Hazard Category H300-H315-H319-H335
Hazard Statement GHS H Statement
H300-H315-H319-H335
Fatal if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 210-436-5
RTECSNumber SK8010000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.