Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pyrogallol, ACS reagent
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 131.0°C to 135.0°C |
Color | White |
Boiling Point | 309.0°C |
Ignition Residue | 0.005% max. |
Infrared Spectrum | Authentic |
Packaging | Plastic bottle |
Linear Formula | C6H3(OH)3 |
Quantity | 100 g |
UN Number | 2811 |
Beilstein | 06, 1071 |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement Harmful to aquatic life with long lasting effects. Toxic if swallowed. Suspected of causing genetic defects. Harmful if inhaled. Harmful in contact with skin. Causes skin irritation. Causes serio |
Precautionary Statement | GHS P Statement IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. If eye irr |
EINECSNumber | 201-762-9 |
RTECSNumber | UX2800000 |
TSCA | TSCA |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction