Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4'-Dibromopropiophenone, 98%, Thermo Scientific™

Catalog No. p-3211511
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle

Chemical Identifiers

CAS 38786-67-3
Molecular Formula C9H8Br2O
Molecular Weight (g/mol) 291.97
MDL Number MFCD00209571
InChI Key GKALOSTUZMFUQB-LURJTMIESA-N
PubChem CID 603907
IUPAC Name 2-bromo-1-(4-bromophenyl)propan-1-one
SMILES C[C@H](Br)C(=O)C1=CC=C(Br)C=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 84°C to 88°C
Color Orange to White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H4COCH(Br)CH3
Quantity 5 g
Synonym 2,4'-dibromopropiophenone, 2-bromo-1-4-bromophenyl propan-1-one, 1-propanone, 2-bromo-1-4-bromophenyl, 2,4-dibromopropiophenone, 2,4'-dibromopropiophenon, acmc-1ago1, alpha,4'-dibromopropiophenone, 1-propanone,2-bromo-1-4-bromophenyl, 2-bromo-1-4-bromophenyl-1-propanone #
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in chloroform,ethylacetate,ethanol,,methanol
Formula Weight 291.97
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2, 4'-Dibromopropiophenone
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.