Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichloro-1-naphthol 97.0+%, TCI America™
SDP

Catalog No. D038325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D038325G 25 g
D03835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D038325G Supplier TCI America Supplier No. D038325G

Chemical Identifiers

CAS 2050-76-2
Molecular Formula C10H6Cl2O
Molecular Weight (g/mol) 213.06
MDL Number MFCD00003931
InChI Key HVLJEMXDXOTWLV-UHFFFAOYSA-N
PubChem CID 16313
IUPAC Name 2,4-dichloronaphthalen-1-ol
SMILES OC1=C2C=CC=CC2=C(Cl)C=C1Cl

Specifications

Melting Point 105°C
Quantity 25 g
Synonym 2,4-dichloro-1-naphthol, 2,4-dichloronaphthol, 1-naphthalenol, 2,4-dichloro, 1-naphthalenol,2,4-dichloro, 2,4-dichloro-.alpha.-naphthol, 1-naphthol, 2,4-dichloro, aurora ka-5007, 1-naphthol,4-dichloro, acmc-1cdm5, 2,4-dichloronaphth-1-ol
Formula Weight 213.06
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dichloro-1-naphthol [for photography]
EINECSNumber (4)-0382
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.