Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichloro-3-cyanopyridine, 95%

Catalog No. AAH63347MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63347MD 250 mg
AAH6334703 1 g
AAH6334706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH63347MD Supplier Thermo Scientific Chemicals Supplier No. H63347MD

CAS: 180995-12-4 | C6H2Cl2N2 | 172.996 g/mol

2,4-Dichloro-3-cyanopyridine, is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Dichloro-3-cyanopyridine, is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 180995-12-4
Molecular Formula C6H2Cl2N2
Molecular Weight (g/mol) 172.996
MDL Number MFCD09750302
InChI Key OQYZEFBWEXEOPL-UHFFFAOYSA-N
PubChem CID 21997828
IUPAC Name 2,4-dichloropyridine-3-carbonitrile
SMILES C1=CN=C(C(=C1Cl)C#N)Cl

Specifications

Quantity 250 mg
UN Number UN3439
Synonym 2,4-dichloro-3-cyanopyridine, 2,4-dichloronicotinonitrile, 2,4-dichloro-3-pyridinecarbonitrile, 3-pyridinecarbonitrile, 2,4-dichloro, 3-cyano-2,4-dichloropyridine, pubchem5457, bys83, 3-cyano-2,4dichloropyridine, ksc493a5f, 3-pyridinecarbonitrile,2,4-dichloro
Solubility Information Slightly soluble in water.
Formula Weight 173
Percent Purity 95%
Chemical Name or Material 2,4-Dichloro-3-cyanopyridine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.