Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichloro-5-fluorobenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. D238625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D238625G 25 g
D23865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D238625G Supplier TCI America Supplier No. D238625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 86522-89-6
Molecular Formula C7H2Cl2FO2
Molecular Weight (g/mol) 207.99
MDL Number MFCD00075281
InChI Key KZCWJHUTTSVCRO-UHFFFAOYSA-M
Synonym 2,4-dichloro-5-fluoro-benzoic acid, 2,4-dichloro-5-fluoro benzoic acid, benzoic acid, 2,4-dichloro-5-fluoro, pubchem1338, maybridge1_002392, acmc-209qa7, ksc448a5p, rarechem al be 1219, timtec-bb sbb003459, 2 4-dichloro-5-fluorobenzoic acid
PubChem CID 688138
IUPAC Name 2,4-dichloro-5-fluorobenzoate
SMILES [O-]C(=O)C1=CC(F)=C(Cl)C=C1Cl

Specifications

Melting Point 144°C
Color White-Yellow
Quantity 25 g
Formula Weight 209.00
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dichloro-5-fluorobenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.