Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichloro-6-methylphenol, 98%

Catalog No. AAH6431203
Click to view available options
Quantity:
250 mg
1 g
5 g

CAS: 1570-65-6 | C7H6Cl2O | 177.024 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1570-65-6
Molecular Formula C7H6Cl2O
Molecular Weight (g/mol) 177.024
MDL Number MFCD00019988
InChI Key WJQZZLQMLJPKQH-UHFFFAOYSA-N
PubChem CID 15292
IUPAC Name 2,4-dichloro-6-methylphenol
SMILES CC1=CC(=CC(=C1O)Cl)Cl

Specifications

Melting Point 52°C to 56°C
Color Brown
Boiling Point 225°C (literature)
Quantity 1 g
UN Number UN3077
Beilstein 1864865
Synonym 4,6-dichloro-o-cresol, 4,6-dichloro-2-methylphenol, phenol, 2,4-dichloro-6-methyl, o-cresol, 4,6-dichloro, 4,6-dichlorocresol, phenol,2,4-dichloro-6-methyl, dichlorocresol, o-cresol,6-dichloro, 4,6-dichlor-o-kresol, acmc-1c0mk
Formula Weight 177.02
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2,4-Dichloro-6-methylphenol
Show More Show Less
Hazard Category H302-H315-H318-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 9; Packaging Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 216-382-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.