Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichloro-6-phenyl-1,3,5-triazine 98.0+%, TCI America™
SDP

Catalog No. D48051G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D48051G 1 g
D48055G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D48051G Supplier TCI America Supplier No. D48051G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1700-02-3
Molecular Formula C9H5Cl2N3
Molecular Weight (g/mol) 226.06
MDL Number MFCD00047340
InChI Key AMEVJOWOWQPPJQ-UHFFFAOYSA-N
PubChem CID 15549
IUPAC Name 2,4-dichloro-6-phenyl-1,3,5-triazine
SMILES C1=CC=C(C=C1)C2=NC(=NC(=N2)Cl)Cl

Specifications

Melting Point 121°C
Color White-Yellow
Boiling Point 136°C
Quantity 1 g
Formula Weight 226.06
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dichloro-6-phenyl-1,3,5-triazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.