Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4'-Dichloroacetophenone, 98+%, Thermo Scientific™

Catalog No. AAA1595414
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1595414 25 g
AAA1595422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1595414 Supplier Thermo Scientific Chemicals Supplier No. A1595414

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 937-20-2
Molecular Formula C8H6Cl2O
Molecular Weight (g/mol) 189.035
MDL Number MFCD00018926
InChI Key FWDFNLVLIXAOMX-UHFFFAOYSA-N
Synonym 2,4'-dichloroacetophenone, 2-chloro-1-4-chlorophenyl ethanone, 4-chlorophenacyl chloride, 2-chloro-1-4-chlorophenyl ethan-1-one, ethanone, 2-chloro-1-4-chlorophenyl, alpha,4-dichloroacetophenone, p-chlorophenacyl chloride, pubchem3372, 2,4-dichloroacetophenone, acmc-209rn9
PubChem CID 70298
IUPAC Name 2-chloro-1-(4-chlorophenyl)ethanone
SMILES C1=CC(=CC=C1C(=O)CCl)Cl

Specifications

Melting Point 99°C to 103°C
Boiling Point 270°C
Quantity 25 g
Beilstein 637861
Formula Weight 189.04
Percent Purity ≥98%
Chemical Name or Material 2,4'-Dichloroacetophenone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H318-H317-H335-H315
Causes serious eye damage.
May cause an allergic skin reaction.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 213-323-9
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.