Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium chloride dihydrate, 99+%, for analysis

Catalog No. AC207780100
Click to view available options
Quantity:
1 kg
2.5 kg
10 kg
Packaging:
Plastic Bottle
Plastic drum

CAS: 10035-04-8 | CaCl2·2H2O | 147.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Adsorbent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10035-04-8
Molecular Formula CaCl2·2H2O
Molecular Weight (g/mol) 147.02

Specifications

Melting Point 175.0°C
Color White
pH 4.5 to 6.5 (5% soln. at 20°C)
Physical Form Crystalline Powder and/or Crystals
Quantity 10 kg
Assay Percent Range 99% min. (Complexometry)
Linear Formula CaCl2·2H2O
Fieser 01,104
Merck Index 15,1661
Solubility Information Solubility in water: 1000g/L (0°C). Other solubilities: freely soluble in alcohol
Formula Weight 147.02
Percent Purity ≥99%
Grade Analytical
Packaging Plastic drum
Specific Gravity 0.83
Density 0.8300g/mL
Chemical Name or Material Calcium chloride dihydrate
Show More Show Less
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.