Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichlorobenzonitrile, 98%

Catalog No. p-7080516
Click to view available options
Quantity:
5 g
25 g

CAS: 6574-98-7 | C7H3Cl2N | 172.008 g/mol

Used as intermediate for the synthesis of agrochemicals, pharmaceuticals and chemical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as intermediate for the synthesis of agrochemicals, pharmaceuticals and chemical intermediates.

Solubility
Soluble in Methanol. Slightly soluble in water.

Notes
Store in cool, dry conditions in well sealed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6574-98-7
Molecular Formula C7H3Cl2N
Molecular Weight (g/mol) 172.008
MDL Number MFCD00016373
InChI Key GRUHREVRSOOQJG-UHFFFAOYSA-N
Synonym benzonitrile, 2,4-dichloro, 2,4-dichloro-benzonitrile, 2,4-dichlorobenzenecarbonitrile, acmc-209nrx, 2,4-dichloro benzonitrile, ksc352s3f
PubChem CID 81050
IUPAC Name 2,4-dichlorobenzonitrile
SMILES C1=CC(=C(C=C1Cl)Cl)C#N

Specifications

Melting Point 57°C to 61°C
Quantity 5 g
UN Number UN3439
Beilstein 637738
Solubility Information Soluble in Methanol. Slightly soluble in water.
Formula Weight 172.01
Percent Purity 98%
Chemical Name or Material 2,4-Dichlorobenzonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 229-493-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.