Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichlorobenzyl Alcohol, Reagent, Spectrum™ Chemical
SDP

Catalog No. 18620660
Click to view available options
Quantity:
25 g
100 g
25 kg

Synonym: (2,4-Dichlorophenyl)methanol, Dybenal, 2,4-Dichlorobenzenemethanol

Chemical Identifiers

CAS 1777-82-8
Molecular Formula C7H6Cl2O
Molecular Weight (g/mol) 177.02
InChI Key DBHODFSFBXJZNY-UHFFFAOYSA-N
IUPAC Name (2,4-dichlorophenyl)methanol
SMILES OCC1=CC=C(Cl)C=C1Cl

Specifications

Quantity 100 g
Grade Reagent
Physical Form Crystalline Powder or Lumps
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.