Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichlorobenzyl chloride, 98%

Catalog No. p-7081452
Click to view available options
Quantity:
500 g
1000 g

CAS: 94-99-5 | C7H5Cl3 | 195.47 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94-99-5
Molecular Formula C7H5Cl3
Molecular Weight (g/mol) 195.47
MDL Number MFCD00000895
InChI Key IRSVDHPYXFLLDS-UHFFFAOYSA-N
Synonym 2,4-dichlorobenzyl chloride, 2,4-dichloro-1-chloromethyl benzene, 2,4-dichlorobenzylchloride, alpha,2,4-trichlorotoluene, benzene, 2,4-dichloro-1-chloromethyl, 2,4-dichloro-1-chloromethyl-benzene, alpha2,4-trichlorotoluene, unii-85e96p2s3s, 2,4-dcbc, toluene, .alpha.2,4-trichloro
PubChem CID 7212
IUPAC Name 2,4-dichloro-1-(chloromethyl)benzene
SMILES ClCC1=CC=C(Cl)C=C1Cl

Specifications

Melting Point -3°C
Density 1.408
Boiling Point 248°C
Flash Point 190°C (374°F)
Odor Pungent
Refractive Index 1.576
Quantity 500 g
UN Number UN3265
Beilstein 387220
Formula Weight 195.48
Percent Purity 98%
Chemical Name or Material 2,4-Dichlorobenzyl chloride
Show More Show Less
Hazard Category H302-H314-H332-H335
Hazard Statement GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 202-381-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.