Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichlorobenzyl Chloride 99%, ACROS Organics™

Click to view available options
Quantity:
5mL
100mL
500mL
Packaging:
Glass bottle
94-99-5
C7H5Cl3
195.48
MFCD00000895
IRSVDHPYXFLLDS-UHFFFAOYSA-N
2,4-dichlorobenzyl chloride,2,4-dichloro-1-chloromethyl benzene,2,4-dichlorobenzylchloride,alpha,2,4-trichlorotoluene,benzene, 2,4-dichloro-1-chloromethyl,2,4-dichloro-1-chloromethyl-benzene,alpha2,4-trichlorotoluene,unii-85e96p2s3s,2,4-dcbc,toluene, .alpha.2,4-trichloro
7212
2,4-dichloro-1-(chloromethyl)benzene
C1=CC(=C(C=C1Cl)Cl)CCl
94-99-5
C7H5Cl3
195.48
MFCD00000895
IRSVDHPYXFLLDS-UHFFFAOYSA-N
2,4-dichlorobenzyl chloride,2,4-dichloro-1-chloromethyl benzene,2,4-dichlorobenzylchloride,alpha,2,4-trichlorotoluene,benzene, 2,4-dichloro-1-chloromethyl,2,4-dichloro-1-chloromethyl-benzene,alpha2,4-trichlorotoluene,unii-85e96p2s3s,2,4-dcbc,toluene, .alpha.2,4-trichloro
7212
2,4-dichloro-1-(chloromethyl)benzene
C1=CC(=C(C=C1Cl)Cl)CCl
Viscosity 2.6 mm2/s (20°C)
Melting Point -2.7°C
Refractive Index 1.5750 to 1.5770
Specific Gravity 1.4
Quantity 500mL
Infrared Spectrum Authentic
Density 1.4000g/mL
Packaging Glass bottle
Color Colorless to Yellow
Boiling Point 248.0°C
Flash Point 190°C
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula Cl2C6H3CH2Cl
Solubility Information Solubility in water: hydrolysis by water. Other solubilities: soluble in alcohol,ether and acetone
Formula Weight 195.48
Physical Form Oily Liquid
Percent Purity 99%
Chemical Name or Material 2, 4-Dichlorobenzyl chloride, 99%
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Very toxic to aquatic life with long lasting effects.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 202-381-0

TSCA : TSCA