Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichlorobenzyl chloride, 99%

Catalog No. p-104241
Click to view available options
Quantity:
5 mL
100 mL
500 mL
Packaging:
Glass bottle

CAS: 94-99-5 | C7H5Cl3 | 195.47 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94-99-5
Molecular Formula C7H5Cl3
Molecular Weight (g/mol) 195.47
MDL Number MFCD00000895
InChI Key IRSVDHPYXFLLDS-UHFFFAOYSA-N
Synonym 2,4-dichlorobenzyl chloride, 2,4-dichloro-1-chloromethyl benzene, 2,4-dichlorobenzylchloride, alpha,2,4-trichlorotoluene, benzene, 2,4-dichloro-1-chloromethyl, 2,4-dichloro-1-chloromethyl-benzene, alpha2,4-trichlorotoluene, unii-85e96p2s3s, 2,4-dcbc, toluene, .alpha.2,4-trichloro
PubChem CID 7212
IUPAC Name 2,4-dichloro-1-(chloromethyl)benzene
SMILES ClCC1=CC=C(Cl)C=C1Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -2.7°C
Color Colorless to Yellow
Density 1.4000g/mL
Boiling Point 248.0°C
Flash Point 190°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5750 to 1.5770
Linear Formula Cl2C6H3CH2Cl
Quantity 5 mL
Specific Gravity 1.4
Solubility Information Solubility in water: hydrolysis by water. Other solubilities: soluble in alcohol,ether and acetone
Viscosity 2.6 mm2/s (20°C)
Formula Weight 195.48
Percent Purity 99%
Physical Form Oily Liquid
Chemical Name or Material 2, 4-Dichlorobenzyl chloride, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Very toxic to aquatic life with long lasting effects.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 202-381-0
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.