Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichloronicotinic acid, 97%, Thermo Scientific™

Catalog No. ac449830010
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle

Chemical Identifiers

CAS 262423-77-8
Molecular Formula C6H3Cl2NO2
Molecular Weight (g/mol) 192.00
MDL Number MFCD06796292
InChI Key ZFGZNVSNOJPGDV-UHFFFAOYSA-N
Synonym 2,4-dichloronicotinic acid, 2,4-dichloronicotinc acid, 2,4-dichloronicotinicacid, 2,4-dichloro-3-pyridinecarboxylic acid, 2.4-dichloronicotinic acid, 2,4-dichloropyridine-3-carboxylicacid, dichloronicotinic acid;, pubchem17746, 2,4-dichloronicotinic acid;
PubChem CID 10583887
IUPAC Name 2,4-dichloropyridine-3-carboxylic acid
SMILES OC(=O)C1=C(Cl)C=CN=C1Cl

Specifications

Melting Point 151°C to 156°C
Assay Percent Range 97%
Packaging Glass bottle
Quantity 1 g
Formula Weight 192
Percent Purity 97%
Chemical Name or Material 2, 4-Dichloronicotinic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.