Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichlorophenyl isothiocyanate, 95%, Thermo Scientific™

Catalog No. AC305280050
Click to view available options
:
5g; Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 6590-96-1
CAS Min % 94.0
CAS Max % 100.0
Density 1.4100g/mL
Boiling Point 260°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Molecular Formula C7H3Cl2NS
Linear Formula Cl2C6H3NCS
Quantity 5g
MDL Number MFCD00037827
Specific Gravity 1.41
Synonym 2,4-dichlorophenyl isothiocyanate, 2,4-dichlorophenylisothiocyanate, benzene,2,4-dichloro-1-isothiocyanato, benzene, 2,4-dichloro-1-isothiocyanato, 2,4-dichlorobenzenisothiocyanate, acmc-1b3n1, tos-bb-1123, 2,4-dichloro-1-isothiocyanato-benzene, #
Solubility Information Solubility in water: 1g/L water (20°C). Other solubilities: soluble in organic solvents
InChI Key WVBNZZHGECFCSH-UHFFFAOYSA-N
SMILES ClC1=CC=C(N=C=S)C(Cl)=C1
IUPAC Name 2,4-dichloro-1-isothiocyanatobenzene
Molecular Weight (g/mol) 204.07
PubChem CID 81066
Formula Weight 204.08
Percent Purity 95%
Chemical Name or Material 2, 4-Dichlorophenyl isothiocyanate, 95%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
EINECSNumber 229-527-6
TSCA TSCA
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.