Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichlorophenylhydrazine hydrochloride, 98%, Thermo Scientific™

Catalog No. AAA1094318
Click to view available options
Quantity:
10 g
50 g
250 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5446-18-4
Molecular Formula C6H7Cl3N2
Molecular Weight (g/mol) 213.486
MDL Number MFCD00012929
InChI Key DDWYGJVFURAIJZ-UHFFFAOYSA-N
Synonym 2,4-dichlorophenylhydrazine hydrochloride, 2,4-dichlorophenyl hydrazine hydrochloride, 2,4-dichlorophenyl hydrazine monohydrochloride, 2,4-dichlorophenylhydrazine hcl, 2,4-dichloro-phenyl-hydrazine hydrochloride, 1-2,4-dichlorophenyl hydrazine hydrochloride, hydrazine, 2,4-dichlorophenyl-, monohydrochloride, pubchem7524, acmc-209lgs, ksc491e2h
PubChem CID 2723905
IUPAC Name (2,4-dichlorophenyl)hydrazine;hydrochloride
SMILES C1=CC(=C(C=C1Cl)Cl)NN.Cl

Specifications

Melting Point ∼212°C (decomposition)
Quantity 50 g
Beilstein 3708828
Formula Weight 213.49
Percent Purity 98%
Chemical Name or Material 2,4-Dichlorophenylhydrazine hydrochloride
Hazard Category H301+H311+H331-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 226-659-6
TSCA No
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.