Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
2,4-Dichloropyridine, 98%
                    Click to view available options
                
                
                    
                            
                                Quantity:
                            
                        
                        
                            
                                5 g
                            
                        
                            
                                25 g
                            
                        
                            
                                100 g
                            
                        
                    Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
 
            Specifications
Specifications
| CAS Min % | 97.5 | 
| CAS Max % | 100.0 | 
| Melting Point | -1.0°C | 
| Density | 1.3700g/mL | 
| Boiling Point | 189.0°C to 190.0°C | 
| Flash Point | 89°C | 
| Assay Percent Range | 97.5% min. (GC) | 
| Packaging | Glass bottle | 
| Refractive Index | 1.5500 to 1.5540 | 
| Quantity | 5 g | 
| Show More | 
Safety and Handling
| Hazard Category | GHS Signal Word: Warning | 
| Hazard Statement | GHS H Statement Causes serious eye irritation. Harmful if inhaled. Causes skin irritation. Harmful if swallowed. May cause respiratory irritation. | 
| Precautionary Statement | GHS P Statement IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse | 
| EINECSNumber | 247-717-7 | 
RUO – Research Use Only
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
             
                                