Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Difluorobenzoyl chloride, 97%, Thermo Scientific™

Catalog No. AC297220025
Click to view available options
:
2.5mL; Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 72482-64-5
CAS Min % 96.0
CAS Max % 100.0
Density 1.42g/mL
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C7H3ClF2O
Refractive Index 1.5147 to 1.5167 (20°C, 589nm)
Linear Formula F2C6H3COCl
Quantity 2.5mL
MDL Number MFCD00000658
Synonym 2,4-difluorobenzoylchloride, benzoyl chloride, 2,4-difluoro, benzoyl chloride,2,4-difluoro, acmc-1bfet, 2,4difluorobenzoylchloride, 2,4-difluorobezoyl chloride, 2,4 difluorobenzoyl chloride, 2,4-difluoro benzoylchloride, 2,4-difluoro-benzoylchloride, 2,4-difluoro-benzoyl chloride
InChI Key JSWRVDNTKPAJLB-UHFFFAOYSA-N
SMILES FC1=CC=C(C(Cl)=O)C(F)=C1
IUPAC Name 2,4-difluorobenzoyl chloride
Molecular Weight (g/mol) 176.55
PubChem CID 588081
Formula Weight 176.55
Percent Purity ≥96.0%
Chemical Name or Material 2, 4-Difluorobenzoyl chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.