Alfa Aesar™ 2,4-Difluorophenyl isocyanate, 98+%

Manufacturer: Alfa Aesar™ A1241522

Quantity 100g

 View more versions of this product

Catalog No. AAA1241522

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 59025-55-7
    Molecular Formula C7H3F2NO
    Formula Weight 155.11
    MDL Number MFCD00001997
    UN Number UN3080
    Beilstein 2831577
    Chemical Name or Material 2,4-Difluorophenyl isocyanate

    Specifications

    Formula Weight 155.11
    Health Hazard 2 GHS H Statement
    H300-H330-H334-H226-H315-H319-H335
    Fatal if swallowed.
    Fatal if inhaled.
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    Flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P210-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 59025-55-7
    Beilstein 2831577
    Health Hazard 1 Danger
    Boiling Point 64° to 66°C (10mmHg)
    Quantity 100g
    Molecular Formula C7H3F2NO
    MDL Number MFCD00001997
    UN Number UN3080
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ISOCYANATES, TOXIC, FLAMMABLE, N.O.S.
    Flash Point 50°C (122°F)
    Chemical Name or Material 2,4-Difluorophenyl isocyanate
    Assay ≥98%

    GHS H Statement
    H300-H330-H334-H226-H315-H319-H335
    Fatal if swallowed.
    Fatal if inhaled.
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    Flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit