Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Difluorotoluene, 98+%

Catalog No. p-7081620
Click to view available options
Quantity:
5 g
25 g

CAS: 452-76-6 | C7H6F2 | 128.122 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 452-76-6
Molecular Formula C7H6F2
Molecular Weight (g/mol) 128.122
MDL Number MFCD00000332
InChI Key MPXDAIBTYWGBSL-UHFFFAOYSA-N
PubChem CID 67983
IUPAC Name 2,4-difluoro-1-methylbenzene
SMILES CC1=C(C=C(C=C1)F)F

Specifications

Density 1.121
Boiling Point 114°C to 115°C
Flash Point 15°C (59°F)
Odor Aromatic
Refractive Index 1.449
Quantity 5 g
UN Number UN2388
Beilstein 1931681
Synonym 2,4-difluorotoluene, difluorotoluene, benzene, 2,4-difluoro-1-methyl, 2,4-difluorotolune, 2, 4-difluorotoluene, 2,4-difluoro methyl benzene, 2,4-difluoro-1-methyl-benzene, 2,4-difluor-1-methylbenzol, pubchem1612, pubchem1613
Formula Weight 128.12
Percent Purity ≥98%
Chemical Name or Material 2,4-Difluorotoluene
Show More Show Less
Hazard Category H225
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLUOROTOLUENES
EINECSNumber 207-211-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.