Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dihydroxybenzamide 98.0+%, TCI America™
SDP

Catalog No. D346025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D346025G 25 g
D34605G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D346025G Supplier TCI America Supplier No. D346025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3147-45-3
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.14
MDL Number MFCD00064913
InChI Key IIUJCQYKTGNRHH-UHFFFAOYSA-N
PubChem CID 76601
IUPAC Name 2,4-dihydroxybenzamide
SMILES NC(=O)C1=CC=C(O)C=C1O

Specifications

Melting Point 228°C
Quantity 25 g
Formula Weight 153.14
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dihydroxybenzamide
EINECSNumber (3)-3064
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.