Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dimethoxyaniline, 97%, Thermo Scientific™

Catalog No. p-114231
Click to view available options
Quantity:
100 g
Packaging:
Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2735-04-8
Molecular Weight (g/mol) 153.18
MDL Number MFCD00008371
InChI Key GEQNZVKIDIPGCO-UHFFFAOYSA-N
Synonym benzenamine, 2,4-dimethoxy, aniline, 2,4-dimethoxy, 2,4-dimethoxyphenylamine, unii-i079354gsi, benzenamine, 2,4-dimethoxy-, hydrochloride, 2,4-domethoxyaniline, 2,4-dimethoxy-anilin, 2, 4-dimethoxyaniline, 2,4 dimethoxy aniline, 2,4-dimethoxy aniline
PubChem CID 17652
IUPAC Name 2,4-dimethoxyaniline
SMILES COC1=CC(=C(C=C1)N)OC

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 34°C to 37°C
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C8H11NO2
Linear Formula (CH3O)2C6H3NH2
Quantity 100 g
Beilstein 13, 784
Formula Weight 153.18
Percent Purity ≥96.0%
Physical Form Crystalline Solid
Chemical Name or Material 2, 4-Dimethoxyaniline, 97%
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.
Harmful in contact with skin.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 220-355-7
RTECSNumber BX4200000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.