Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dimethoxybenzaldoxime 95.0+%, TCI America™
SDP

Catalog No. D485125G
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 31874-34-7
Molecular Formula C9H11NO3
Molecular Weight (g/mol) 181.191
MDL Number MFCD00082783
InChI Key SFDRVCQSVTYHLU-UXBLZVDNSA-N
Synonym 2,4-dimethoxybenzaldoxime, 2,4-dimethoxybenzaldehyde oxime, e-n-2,4-dimethoxyphenyl methylidene hydroxylamine, benzaldehyde, 2,4-dimethoxy-, oxime, 2.4-dimethoxybenzaldoxim, e-2,4-dimethoxybenzaldehyde oxime, 2,4-dimethoxyphenyl hydroxyimino methane, ne-n-2,4-dimethoxyphenyl methylidene hydroxylamine
PubChem CID 6871293
IUPAC Name (NE)-N-[(2,4-dimethoxyphenyl)methylidene]hydroxylamine
SMILES COC1=CC(=C(C=C1)C=NO)OC

Specifications

Melting Point 106°C
Color White
Quantity 25 g
Formula Weight 181.19
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dimethoxybenzaldoxime
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.