Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dimethoxyphenylboronic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
133730-34-4
C8H11BO4
181.982
MFCD01074590
SQTUYFKNCCBFRR-UHFFFAOYSA-N
2,4-dimethoxybenzeneboronic acid,2,4-dimethoxyphenyl boronic acid,2,4-dimethoxyphenyl boranediol,boronic acid, 2,4-dimethoxyphenyl,2,4-dimethoxybenzene boronic acid,boronic acid, b-2,4-dimethoxyphenyl,pubchem1823,acmc-209btd,ksc174i6j,2,4-dimethoxyphenylboronicacid
2734341
(2,4-dimethoxyphenyl)boronic acid
B(C1=C(C=C(C=C1)OC)OC)(O)O
133730-34-4
C8H11BO4
181.982
MFCD01074590
SQTUYFKNCCBFRR-UHFFFAOYSA-N
2,4-dimethoxybenzeneboronic acid,2,4-dimethoxyphenyl boronic acid,2,4-dimethoxyphenyl boranediol,boronic acid, 2,4-dimethoxyphenyl,2,4-dimethoxybenzene boronic acid,boronic acid, b-2,4-dimethoxyphenyl,pubchem1823,acmc-209btd,ksc174i6j,2,4-dimethoxyphenylboronicacid
2734341
(2,4-dimethoxyphenyl)boronic acid
B(C1=C(C=C(C=C1)OC)OC)(O)O
Quantity 1g
Color Beige to White or Pink
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 181.98
Physical Form Moist Powder
Percent Purity ≥96.0%
Chemical Name or Material 2, 4-Dimethoxybenzeneboronic acid
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.