Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dimethyl-1,3-pentadiene, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
1000-86-8
C7H12
96.17
MFCD00008903
CMSUNVGIWAFNBG-UHFFFAOYSA-N
2,4-dimethyl-1,3-pentadiene,1,1,3-trimethylbutadiene,1,3-pentadiene, 2,4-dimethyl,acmc-20aoci,1,3-trimethylbutadiene,1,3-pentadiene,2,4-dimethyl,cmsunvgiwafnbg-uhfffaoysa,ch3 2c=chc ch3 =ch2,1,3-pentadiene, 2,4-dimethyl-8ci 9ci
66080
2,4-dimethylpenta-1,3-diene
CC(=CC(=C)C)C
1000-86-8
C7H12
96.17
MFCD00008903
CMSUNVGIWAFNBG-UHFFFAOYSA-N
2,4-dimethyl-1,3-pentadiene,1,1,3-trimethylbutadiene,1,3-pentadiene, 2,4-dimethyl,acmc-20aoci,1,3-trimethylbutadiene,1,3-pentadiene,2,4-dimethyl,cmsunvgiwafnbg-uhfffaoysa,ch3 2c=chc ch3 =ch2,1,3-pentadiene, 2,4-dimethyl-8ci 9ci
66080
2,4-dimethylpenta-1,3-diene
CC(=CC(=C)C)C
Boiling Point 94°C
Flash Point 10°C
Specific Gravity 0.74
Density 0.7400g/mL
Refractive Index 1.4400 to 1.442
Color Colorless to Yellow
Quantity 5g
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula (CH3)2C=CHC(CH3)=CH2
Beilstein 01, 257
Formula Weight 96.17
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 2, 4-Dimethyl-1, 3-pentadiene
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 213-677-4