Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dimethyl-3-pentanol 99.0+%, TCI America™
SDP

Catalog No. D238725ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D238725ML 25 mL
D2387500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D238725ML Supplier TCI America Supplier No. D238725ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 600-36-2
Molecular Formula C7H16O
Molecular Weight (g/mol) 116.204
MDL Number MFCD00008921
InChI Key BAYAKMPRFGNNFW-UHFFFAOYSA-N
Synonym 2,4-dimethyl-3-pentanol, diisopropylcarbinol, diisopropylmethanol, 3-pentanol, 2,4-dimethyl, di-iso-propylcarbinol, 2,4-dimethyl-3-hydroxypentane, unii-n54k81p01f, g00025-watson-int, ethyl tert-butyl carbinol, acmc-1adj1
PubChem CID 11752
IUPAC Name 2,4-dimethylpentan-3-ol
SMILES CC(C)C(C(C)C)O

Specifications

Color Colorless
Boiling Point 136°C
Quantity 25 mL
UN Number 1987
Formula Weight 116.20
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4-Dimethyl-3-pentanol
EINECSNumber (2)-0217
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.