Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dimethylaniline, 99%, ACROS Organics™

Click to view available options
Quantity:
100g
1kg
Packaging:
Glass bottle
95-68-1
C8H11N
121.18
MFCD00007738
CZZZABOKJQXEBO-UHFFFAOYSA-N
2,4-xylidine,2,4-dimethyl aniline,m-xylidine,m-4-xylidine,2,4-dimethylphenylamine,benzenamine, 2,4-dimethyl,2-methyl-p-toluidine,4-methyl-o-toluidine,1-amino-2,4-dimethylbenzene,4-amino-1,3-xylene
7250
CHEBI:27840
2,4-dimethylaniline
CC1=CC(=C(C=C1)N)C
95-68-1
C8H11N
121.18
MFCD00007738
CZZZABOKJQXEBO-UHFFFAOYSA-N
2,4-xylidine,2,4-dimethyl aniline,m-xylidine,m-4-xylidine,2,4-dimethylphenylamine,benzenamine, 2,4-dimethyl,2-methyl-p-toluidine,4-methyl-o-toluidine,1-amino-2,4-dimethylbenzene,4-amino-1,3-xylene
7250
CHEBI:27840
2,4-dimethylaniline
CC1=CC(=C(C=C1)N)C
Specific Gravity 0.98
Refractive Index 1.5580 to 1.5600
Quantity 100g
Flash Point 90°C
Packaging Glass bottle
Infrared Spectrum Authentic
Boiling Point 218.0°C
Density 0.9800g/mL
Melting Point 16.0°C
Color Undesignated
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Linear Formula (CH3)2C6H3NH2
Beilstein 12, 1111
Merck Index 15, 10282
Solubility Information Solubility in water: 5g/L (20°C). Other solubilities: soluble in ethanol, ether and benzene
Formula Weight 121.18
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 2,4-Dimethylaniline, 99%
Show More Show Less

GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Toxic to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER o

GHS Signal Word: Danger

EINECSNumber : 202-440-0

RTECSNumber : ZE8925000

TSCA : TSCA

Recommended Storage : Product darkens if exposed to light and air