Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dimethylaniline, 99%

Catalog No. p-115366
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Glass bottle

CAS: 95-68-1 | C8H11N | 121.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-68-1
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.18
MDL Number MFCD00007738
InChI Key CZZZABOKJQXEBO-UHFFFAOYSA-N
Synonym 2,4-xylidine, 2,4-dimethyl aniline, m-xylidine, m-4-xylidine, 2,4-dimethylphenylamine, benzenamine, 2,4-dimethyl, 2-methyl-p-toluidine, 4-methyl-o-toluidine, 1-amino-2,4-dimethylbenzene, 4-amino-1,3-xylene
PubChem CID 7250
ChEBI CHEBI:27840
IUPAC Name 2,4-dimethylaniline
SMILES CC1=CC(=C(C=C1)N)C

Specifications

Melting Point 16.0°C
CAS Min % 98.5
CAS Max % 100.0
Density 0.9800g/mL
Boiling Point 218.0°C
Quantity 100 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.5580 to 1.5600
Assay Percent Range 98.5% min. (GC)
Specific Gravity 0.98
Linear Formula (CH3)2C6H3NH2
Beilstein 12, 1111
Merck Index 15, 10282
Solubility Information Solubility in water: 5g/L (20°C). Other solubilities: soluble in ethanol, ether and benzene
Formula Weight 121.18
Percent Purity 99%
Chemical Name or Material 2,4-Dimethylaniline, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER o
EINECSNumber 202-440-0
RTECSNumber ZE8925000
TSCA TSCA
Recommended Storage Product darkens if exposed to light and air

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.