Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dimethylbenzaldehyde, 99%

Catalog No. p-3736303
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle

CAS: 15764-16-6 | C9H10O | 134.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15764-16-6
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.18
MDL Number MFCD00003340
InChI Key GISVICWQYMUPJF-UHFFFAOYSA-N
Synonym benzaldehyde, 2,4-dimethyl, 2,4-dimethyl benzaldehyde, 2,4-xylylaldehyde, 2,4-dimethylbenzenecarboxaldehyde, unii-i06yu18h4a, 1-formyl-2,4-dimethylbenzene, 2,4-dimethyl-benzaldehyde, m-xylylaldehyde, fema no. 3427, 2,4-trimethyl benzaldehyde
PubChem CID 61814
IUPAC Name 2,4-dimethylbenzaldehyde
SMILES CC1=CC(=C(C=C1)C=O)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -9°C
Color Colorless
Density 0.9600g/mL
Boiling Point 102.4°C to 103°C (14.0 mmHg)
Flash Point 88°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5482 to 1.5502
Linear Formula (CH3)2C6H3CHO
Quantity 5 g
Beilstein 07, IV, 706
Specific Gravity 0.96
Formula Weight 134.18
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2, 4-Dimethylbenzaldehyde
Show More Show Less
EINECSNumber 239-856-7

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.