Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dimethylbenzenesulfonic Acid Sodium Salt 95%, Thermo Scientific™

Catalog No. AC408200250
Click to view available options
Quantity:
25 g
Packaging:
Glass Bottle

Chemical Identifiers

CAS 827-21-4
Molecular Formula C8H9NaO3S
Molecular Weight (g/mol) 208.207
MDL Number MFCD00041883
InChI Key GJQXYSKWRDJNAJ-UHFFFAOYSA-M
Synonym sodium 2,4-dimethylbenzenesulfonate, sodium m-xylene-4-sulfonate, m-xylene-4-sulfonic acid sodium salt, benzenesulfonic acid, 2,4-dimethyl-, sodium salt, benzenesulfonic acid, 2,4-dimethyl-, sodium salt 1:1, 2,4-dimethylbenzenesulfonic acid sodium salt, sodium m-xylene-4-sulphonate, acmc-20ajps, dsstox_cid_21639
PubChem CID 23677979
IUPAC Name sodium;2,4-dimethylbenzenesulfonate
SMILES CC1=CC(=C(C=C1)S(=O)(=O)[O-])C.[Na+]

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass Bottle
Quantity 25 g
Formula Weight 208.21
Percent Purity ≥94%
Physical Form Powder
Chemical Name or Material 2, 4-Dimethylbenzenesulfonic acid sodium salt, 95%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.