Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dimethylnitrobenzene 98.0+%, TCI America™
SDP

Catalog No. p-7126116
Click to view available options
Quantity:
25 g
500 g

Chemical Identifiers

CAS 89-87-2
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD00007169
InChI Key BBUPBICWUURTNP-UHFFFAOYSA-N
Synonym 4-nitro-m-xylene, 1,3-dimethyl-4-nitrobenzene, 4-nitro-1,3-dimethylbenzene, 2,4-dimethylnitrobenzene, benzene, 2,4-dimethyl-1-nitro, m-xylene, 4-nitro, 4-nitro-1,3-xylene, 1-nitro-2,4-dimethylbenzene, unii-7em84eue05, ccris 3120
PubChem CID 6991
IUPAC Name 2,4-dimethyl-1-nitrobenzene
SMILES CC1=CC(=C(C=C1)[N+](=O)[O-])C

Specifications

Color Yellow
Boiling Point 245°C
Quantity 25 g
UN Number 1665
Formula Weight 151.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4-Dimethylnitrobenzene
EINECSNumber (3)-0399
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.