Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dimethylphenol, 99%

Catalog No. ac408450050
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle

CAS: 105-67-9 | C8H10O | 122.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-67-9
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.17
MDL Number MFCD00002233
InChI Key KUFFULVDNCHOFZ-UHFFFAOYSA-N
Synonym 2,4-xylenol, phenol, 2,4-dimethyl, gallex, 4,6-dimethylphenol, 1-hydroxy-2,4-dimethylbenzene, 4-hydroxy-1,3-dimethylbenzene, rcra waste number u101, 2,4-dimethyl phenol, caswell no. 907a, lysol brand disinfectant
PubChem CID 7771
ChEBI CHEBI:34241
IUPAC Name 2,4-dimethylphenol
SMILES CC1=CC(=C(C=C1)O)C

Specifications

Melting Point 21.0°C to 23.0°C
Color Colorless to Yellow
CAS Min % 98.5
CAS Max % 100.0
Density 1.0270g/mL
Boiling Point 211.0°C
Quantity 5 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.5375 to 1.5405
Assay Percent Range 98.5% min. (GC)
Specific Gravity 1.027
Linear Formula (CH3)2C6H3OH
Beilstein 06, 486
Merck Index 15, 10279
Solubility Information Solubility in water: 0.5% (25°C). Other solubilities: freely soluble in alcohol,chloroform,ether,benz,ene,soluble in naoh solutions
Formula Weight 122.17
Percent Purity 99%
Chemical Name or Material 2, 4-Dimethylphenol
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Gently wash with plenty of soap and water.
IF IN EYES: Rinse cautiously wit
EINECSNumber 203-321-6

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.