Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Antimony pentasulfide, pure

Catalog No. AC330412500
Click to view available options
Quantity:
50 g
250 g
1 kg

Catalog No. AC330412500

Supplier: Thermo Scientific Chemicals 330412500

Only null left
Add to Cart

CAS: 1315-04-4 | S5Sb2 | 403.82 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1315-04-4
Molecular Formula S5Sb2
Molecular Weight (g/mol) 403.82
MDL Number MFCD00016321
InChI Key PPKVREKQVQREQD-UHFFFAOYSA-N
Synonym antimony pentasulfide, antimony v sulfide, antimony red, antimonic sulfide, antimonial saffron, antimony sulfide sb2s5, antimony sulfide golden, golden antimony sulfide, diantimony pentasulphide, goldschwefel
PubChem CID 16683083
IUPAC Name [bis(sulfanylidene)-$l^{5}-stibanyl]sulfanyl-bis(sulfanylidene)-$l^{5}-stibane
SMILES S=[Sb](=S)S[Sb](=S)=S

Specifications

Quantity 250 g
Linear Formula Sb2S5
Merck Index 15,688
Solubility Information Solubility in water: insoluble.
Formula Weight 403.83
Percent Purity ≥54% (Sb)
Grade Pure
Chemical Name or Material Antimony pentasulfide
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable solid.
Harmful if swallowed.
Harmful in contact with skin.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED
EINECSNumber 215-255-5

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.