Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dinitroaniline, 99%

Catalog No. p-120364
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle

CAS: 97-02-9 | C6H5N3O4 | 183.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-02-9
Molecular Formula C6H5N3O4
Molecular Weight (g/mol) 183.12
MDL Number MFCD00007151
InChI Key LXQOQPGNCGEELI-UHFFFAOYSA-N
Synonym benzenamine, 2,4-dinitro, 1-amino-2,4-dinitrobenzene, 2,4-dinitroanilin, 2,4-dinitrobenzenamine, 2,4-dinitroanilina, 2,4-dinitraniline, 2,4-dinitrophenylamine, aniline, 2,4-dinitro, 2,4-dinitroaminobenzene, 2,4-nitroaniline
PubChem CID 7321
ChEBI CHEBI:34242
IUPAC Name 2,4-dinitroaniline
SMILES NC1=CC=C(C=C1[N+]([O-])=O)[N+]([O-])=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 177°C to 180°C
Color Brown-Yellow or Green-Yellow to Yellow
Flash Point 224°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Linear Formula (O2N)2C6H3NH2
Quantity 5 g
Beilstein 12,747
Merck Index 15,3296
Solubility Information Solubility in water: 0.06g/L (20 c). Other solubilities: 0.7% ethanol (21 c),soluble in diethyl ether
Formula Weight 183.12
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2, 4-Dinitroaniline, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Fatal if inhaled.
Toxic to aquatic life with long lasting effects.
Fatal if swallowed.
Fatal in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 202-553-5
RTECSNumber BX9100000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.