Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dinitroaniline, 99% (dry wt.), cont. up to 15% water

Catalog No. AAB210800E
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB210800E 2500 g
AAB2108022 100 g
AAB2108036 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB210800E Supplier Thermo Scientific Chemicals Supplier No. B210800E
Add to Cart

CAS: 97-02-9 | C6H5N3O4 | 183.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-02-9
Molecular Formula C6H5N3O4
Molecular Weight (g/mol) 183.12
MDL Number MFCD00007151
InChI Key LXQOQPGNCGEELI-UHFFFAOYSA-N
Synonym benzenamine, 2,4-dinitro, 1-amino-2,4-dinitrobenzene, 2,4-dinitroanilin, 2,4-dinitrobenzenamine, 2,4-dinitroanilina, 2,4-dinitraniline, 2,4-dinitrophenylamine, aniline, 2,4-dinitro, 2,4-dinitroaminobenzene, 2,4-nitroaniline
PubChem CID 7321
ChEBI CHEBI:34242
IUPAC Name 2,4-dinitroaniline
SMILES NC1=CC=C(C=C1[N+]([O-])=O)[N+]([O-])=O

Specifications

Melting Point 176°C to 180°C
Density 1.61
Flash Point 224°C (435°F)
Quantity 2500 g
UN Number UN1596
Beilstein 982999
Merck Index 14,3270
Formula Weight 183.12
Percent Purity 99%
Assay (dry wt.), cont. up to 15% water
Chemical Name or Material 2,4-Dinitroaniline
Show More Show Less
Hazard Category H300+H310+H330-H373
Hazard Statement GHS H Statement
H310-H330-H373-H302
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Precautionary Statement P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P314-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: DINITROANILINES
EINECSNumber 202-553-5
RTECSNumber BX9100000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.