Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dinitrophenylacetic acid, 98%

Catalog No. AAB2210222
Click to view available options
Quantity:
25 g
100 g

CAS: 643-43-6 | C8H6N2O6 | 226.144 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 643-43-6
Molecular Formula C8H6N2O6
Molecular Weight (g/mol) 226.144
MDL Number MFCD00007227
InChI Key KCNISYPADDTFDO-UHFFFAOYSA-N
Synonym 2,4-dinitrophenylacetic acid, 2-2,4-dinitrophenyl acetic acid, 2,4-dinitrophenyl acetic acid, benzeneacetic acid, 2,4-dinitro, acmc-209nlg, 2,4-dimtrophenylacetic acid, ncistruc1_000234, ncistruc2_000110, 2,4-dinitrobenzeneacetic acid, benzeneacetic acid,4-dinitro
PubChem CID 221513
SMILES C1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])CC(=O)O

Specifications

Melting Point 177°C to 182°C
Quantity 100 g
Beilstein 1990798
Formula Weight 226.15
Percent Purity 98%
Chemical Name or Material 2,4-Dinitrophenylacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 211-398-2
RTECSNumber AH2230000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.