Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dinitrophenylhydrazine, Spectrum™

Click to view available options
Quantity:
25 g
100 g
500 g
10 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
119-26-6
C6H6N4O4
198.14
HORQAOAYAYGIBM-UHFFFAOYSA-N
(2,4-dinitrophenyl)hydrazine
NNC1=CC=C(C=C1[N+]([O-])=O)[N+]([O-])=O
119-26-6
C6H6N4O4
198.14
HORQAOAYAYGIBM-UHFFFAOYSA-N
(2,4-dinitrophenyl)hydrazine
NNC1=CC=C(C=C1[N+]([O-])=O)[N+]([O-])=O

DI149, 119-26-6, C6H6N4O4

Spectrum™ 2,4-Dinitrophenylhydrazine is a chemical compound that is usually supplied as a wet red-to-orange solid that is shock sensitive. It is used in Brady′s test to detect the carbonyl functionality of an aldehyde or ketone functional group.
Water 30%
Packaging Amber Glass Bottle
Quantity 25 g
CAS Max % 100%
UN Number UN1325