Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-(4-Fluorophenyl)benzimidazole, 95%

Click to view available options
Quantity:
250mg
1g
324-27-6
C13H9FN2
212.227
MFCD00224358
FPWUSPPQEHBWHC-UHFFFAOYSA-N
2-4-fluorophenyl-1h-benzimidazole,1h-benzimidazole, 2-4-fluorophenyl,2-4-fluorophenyl-1h-1,3-benzodiazole,2-4-fluorophenyl-1h-benzoimidazole,p-fluorophenyl-2 benzimidazole,2-4-fluoro-phenyl-1h-benzoimidazole,2-4-fluorophenyl-1h-benzo d imidazole,p-fluorophenyl-2 benzimidazole french,2-4-fluorophenyl benzimidazole,benzimidazole, 2-p-fluorophenyl-7ci,8ci
101259
2-(4-fluorophenyl)-1H-benzimidazole
C1=CC=C2C(=C1)NC(=N2)C3=CC=C(C=C3)F
324-27-6
C13H9FN2
212.227
MFCD00224358
FPWUSPPQEHBWHC-UHFFFAOYSA-N
2-4-fluorophenyl-1h-benzimidazole,1h-benzimidazole, 2-4-fluorophenyl,2-4-fluorophenyl-1h-1,3-benzodiazole,2-4-fluorophenyl-1h-benzoimidazole,p-fluorophenyl-2 benzimidazole,2-4-fluoro-phenyl-1h-benzoimidazole,2-4-fluorophenyl-1h-benzo d imidazole,p-fluorophenyl-2 benzimidazole french,2-4-fluorophenyl benzimidazole,benzimidazole, 2-p-fluorophenyl-7ci,8ci
101259
2-(4-fluorophenyl)-1H-benzimidazole
C1=CC=C2C(=C1)NC(=N2)C3=CC=C(C=C3)F
Quantity 1g
Formula Weight 212.23
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 2-(4-Fluorophenyl)benzimidazole

RTECSNumber : DD8969232