Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Hexadiene, 90%, Tech., mixture of isomers, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
592-46-1
C6H10
82.14
MFCD00009297
APPOKADJQUIAHP-CIIODKQPSA-N
unii-g4m9133vs9,2,4-hexadiene, e,z,cis,trans-2,4-hexadiene,2,4-hexadiene,e , z-ch3ch=chch=chch3,cis,trans-hexa-2,4-diene,2,4-hexadiene, 2e,4z,cis-2-trans-4-hexadiene,e,z-2,4-hexadiene,z,e-2,4-hexadiene
643786
(2Z,4E)-hexa-2,4-diene
CC=CC=CC
592-46-1
C6H10
82.14
MFCD00009297
APPOKADJQUIAHP-CIIODKQPSA-N
unii-g4m9133vs9,2,4-hexadiene, e,z,cis,trans-2,4-hexadiene,2,4-hexadiene,e , z-ch3ch=chch=chch3,cis,trans-hexa-2,4-diene,2,4-hexadiene, 2e,4z,cis-2-trans-4-hexadiene,e,z-2,4-hexadiene,z,e-2,4-hexadiene
643786
(2Z,4E)-hexa-2,4-diene
CC=CC=CC
Flash Point -8°C
Boiling Point 82°C
Refractive Index 1.45
Specific Gravity 0.72
Density 0.7200g/mL
Infrared Spectrum Authentic
Packaging Glass bottle
Color Yellow
Quantity 1g
CAS Min % 88.0
CAS Max % 100.0
Assay Percent Range 88% min. (GC)
Linear Formula CH3CH=CHCH=CHCH3
Beilstein 01, 254
Solubility Information Solubility in water: insoluble
Formula Weight 82.14
Physical Form Liquid
Percent Purity 90%
Grade Technical
Chemical Name or Material 2, 4-Hexadiene, tech. 90%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 209-757-3

TSCA : TSCA